C-[1-(3-Fluorophenyl)cyclopentyl]methylamine

Supplier: Matrix Scientific

021300-500MG
101811-730EA 370.3 USD
101811-730
C-[1-(3-Fluorophenyl)cyclopentyl]methylamine
C-[1-(3-Fluorophenyl)cyclopentyl]methylamine

Hazard: Irritant

Formula: C₁₂H₁₆FN
MDL Number: MFCD06213186
CAS Number: 359715-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR