3-(Piperidin-4-yloxy)pyridine

Supplier: Matrix Scientific

021297-500MG
101811-724EA 179.4 USD
101811-724
3-(Piperidin-4-yloxy)pyridine
3-(Piperidin-4-yloxy)pyridine

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
MW: 178.24 g/mol
MDL Number: MFCD08059774
CAS Number: 310881-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR