4-[4-(2-Hydroxyethyl)piperazin-1-yl]benzoic acid

Supplier: Matrix Scientific

021292-500MG
101811-714EA 209.3 USD
101811-714
4-[4-(2-Hydroxyethyl)piperazin-1-yl]benzoic acid
4-[4-(2-Hydroxyethyl)piperazin-1-yl]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD06742259
CAS Number: 889939-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR