3-(3-Fluorophenoxy)propionic acid

Supplier: Matrix Scientific

Synonyms: 3-(3-Fluorophenoxy)propionic Acid

021279-500MG
101811-692EA 209.3 USD
101811-692
3-(3-Fluorophenoxy)propionic acid
3-(3-Fluorophenoxy)propionic acid

Hazard: Irritant

Formula: C₉H₉FO₃
MW: 184.1644 g/mol
MDL Number: MFCD01931913
CAS Number: 133077-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR