4-Ethyl[1,2,3]thiadiazole-5-carbonyl chloride

Supplier: Matrix Scientific

021258-500MG
101811-646EA 209.3 USD
101811-646
4-Ethyl[1,2,3]thiadiazole-5-carbonyl chloride
4-Ethyl[1,2,3]thiadiazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₅H₅ClN₂OS
MDL Number: MFCD08445752
CAS Number: 889939-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR