L(-)-Methioninol

Supplier: Matrix Scientific

Synonyms: (S)-2-Amino-4-(methylthio)butan-1-olL-Methioninol

021255-500MG
101811-640EA 280 USD
101811-640
L(-)-Methioninol
L(-)-Methioninol

Hazard: Irritant

Formula: C₅H₁₃NOS
MW: 135.2 g/mol
MDL Number: MFCD00004735
CAS Number: 2899-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR