(1R,2S)-2-Methylamino-cyclohexanol

Supplier: Matrix Scientific

021248-500MG
101811-626EA 280 USD
101811-626
(1R,2S)-2-Methylamino-cyclohexanol
(1R,2S)-2-Methylamino-cyclohexanol

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD08445777
CAS Number: 20431-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR