1-(3-Fluoro-4-methoxyphenyl)ethylamine

Supplier: Matrix Scientific

Synonyms: 1-(3-Fluoro-4-methoxyphenyl)ethanamine

021234-500MG
101811-600EA 209.3 USD
101811-600
1-(3-Fluoro-4-methoxyphenyl)ethylamine
1-(3-Fluoro-4-methoxyphenyl)ethylamine

Hazard: Irritant

Formula: C₉H₁₂FNO
MW: 169.2 g/mol
MDL Number: MFCD04116349
CAS Number: 105321-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR