1-Thiazol-2-ylpiperazine

Supplier: Matrix Scientific

Synonyms: 2-(1-Piperazinyl)thiazole1-(2-Thiazolyl)piperazine

021228-500MG
101811-588EA 195.5 USD
101811-588
1-Thiazol-2-ylpiperazine
1-Thiazol-2-ylpiperazine

Hazard: Irritant

Formula: C₇H₁₁N₃S
MW: 169.25 g/mol
MDL Number: MFCD00778419
CAS Number: 42270-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR