C-(3,4-Dimethylphenyl)-C-pyridin-4-ylmethylamine

Supplier: Matrix Scientific

021205-500MG
101811-550EA 370.3 USD
101811-550
C-(3,4-Dimethylphenyl)-C-pyridin-4-ylmethylamine
C-(3,4-Dimethylphenyl)-C-pyridin-4-ylmethylamine

Hazard: Irritant

Formula: C₁₄H₁₆N₂
MDL Number: MFCD08445741
CAS Number: 889939-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR