3-Pyrazol-1-ylpropionic acid

Supplier: Matrix Scientific

Synonyms: 3-(1-Pyrazolyl)propionic Acid

021196-500MG
101811-534EA 280 USD
101811-534
3-Pyrazol-1-ylpropionic acid
3-Pyrazol-1-ylpropionic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD01822327
CAS Number: 89532-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR