3-[5-(4-Chlorophenyl)tetrazol-1-yl]propionic acid

Supplier: Matrix Scientific

021169-500MG
101811-492EA 209.3 USD
101811-492
3-[5-(4-Chlorophenyl)tetrazol-1-yl]propionic acid
3-[5-(4-Chlorophenyl)tetrazol-1-yl]propionic acid

Hazard: Irritant

Formula: C₁₀H₉ClN₄O₂
MDL Number: MFCD08445718
CAS Number: 889939-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR