1-[1-(3-Fluorophenyl)ethyl]-1H-[1,2,3]triazole-4-carboxylic acid

Supplier: Matrix Scientific

021168-500MG
101811-490EA 209.3 USD
101811-490
1-[1-(3-Fluorophenyl)ethyl]-1H-[1,2,3]triazole-4-carboxylic acid
1-[1-(3-Fluorophenyl)ethyl]-1H-[1,2,3]triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀FN₃O₂
MDL Number: MFCD08445763
CAS Number: 889939-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR