3-(5-p-Tolyltetrazol-1-yl)propionic acid

Supplier: Matrix Scientific

021167-500MG
101811-488EA 209.3 USD
101811-488
3-(5-p-Tolyltetrazol-1-yl)propionic acid
3-(5-p-Tolyltetrazol-1-yl)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₄O₂
MDL Number: MFCD08445818
CAS Number: 889940-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR