7-Chloro-2,5-dimethyl-3-thiophen-2-ylpyrazolo[1,5-a]pyrimidine

Supplier: Matrix Scientific

021162-500MG
101811-480EA 248.4 USD
101811-480
7-Chloro-2,5-dimethyl-3-thiophen-2-ylpyrazolo[1,5-a]pyrimidine
7-Chloro-2,5-dimethyl-3-thiophen-2-ylpyrazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃S
MDL Number: MFCD08445706
CAS Number: 889939-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR