7,8-Dimethyl-1,2,3,9b-tetraazacyclopenta[a]naphthalene-4-carbaldehyde

Supplier: Matrix Scientific

021157-500MG
101811-470EA 195.5 USD
101811-470
7,8-Dimethyl-1,2,3,9b-tetraazacyclopenta[a]naphthalene-4-carbaldehyde
7,8-Dimethyl-1,2,3,9b-tetraazacyclopenta[a]naphthalene-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₀N₄O
MDL Number: MFCD08445742
CAS Number: 889939-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR