2-Fluoro-N-(2-methoxyethyl)-N-[5-(4-methoxyphenyl)[1,3,4]thiadiazol-2-yl]benzamide

Supplier: Matrix Scientific

021155-500MG
101811-466EA 224.25 USD
101811-466
2-Fluoro-N-(2-methoxyethyl)-N-[5-(4-methoxyphenyl)[1,3,4]thiadiazol-2-yl]benzamide
2-Fluoro-N-(2-methoxyethyl)-N-[5-(4-methoxyphenyl)[1,3,4]thiadiazol-2-yl]benzamide

Hazard: Irritant

Formula: C₁₉H₁₈FN₃O₃S
MDL Number: MFCD04200758
CAS Number: 603074-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR