2-(4-Fluorophenoxy)-N-methyl-1-ethanamine hydrochloride

Supplier: Matrix Scientific

021138-500MG
101811-426EA 228.85 USD
101811-426
2-(4-Fluorophenoxy)-N-methyl-1-ethanamine hydrochloride
2-(4-Fluorophenoxy)-N-methyl-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClFNO
MDL Number: MFCD08445756
CAS Number: 1051919-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR