1-(3,4,5-Trimethoxyphenyl)ethylamine

Supplier: Matrix Scientific

021082-500MG
101811-312EA 201.25 USD
101811-312
1-(3,4,5-Trimethoxyphenyl)ethylamine
1-(3,4,5-Trimethoxyphenyl)ethylamine

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD02656660
CAS Number: 121082-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR