N-(3-tert-Butylisoxazol-5-yl)-2-chloroacetamide

Supplier: Matrix Scientific

021022-500MG
101811-202EA 398.46 USD
101811-202
N-(3-tert-Butylisoxazol-5-yl)-2-chloroacetamide
N-(3-tert-Butylisoxazol-5-yl)-2-chloroacetamide

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₂
MW: 216.67 g/mol
MDL Number: MFCD06655789
CAS Number: 668980-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR