ω-Chloro-3,4-difluoroacetophenone

Supplier: Matrix Scientific

Synonyms: 2-Chloro-1-(3,4-difluorophenyl)ethanone2-Chloro-3',4'-difluoroacetophenone

021009-500MG
101811-180EA 209.3 USD
101811-180
ω-Chloro-3,4-difluoroacetophenone
ω-Chloro-3,4-difluoroacetophenone

Hazard: Irritant

Formula: C₈H₅ClF₂O
MW: 190.5745 g/mol
MDL Number: MFCD03966888
CAS Number: 51336-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR