C-(2-Methyl-2H-pyrazol-3-yl)methylamine dihydrochloride

Supplier: Matrix Scientific

021008-500MG
101811-178EA 235.75 USD
101811-178
C-(2-Methyl-2H-pyrazol-3-yl)methylamine dihydrochloride
C-(2-Methyl-2H-pyrazol-3-yl)methylamine dihydrochloride

Hazard: Irritant

Formula: C₅H₁₁Cl₂N₃
MDL Number: MFCD11506366
CAS Number: 1185299-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR