3-[Methyl-(1-methylpiperidin-4-yl)amino]propionic acid

Supplier: Matrix Scientific

021006-500MG
101811-174EA 201.25 USD
101811-174
3-[Methyl-(1-methylpiperidin-4-yl)amino]propionic acid
3-[Methyl-(1-methylpiperidin-4-yl)amino]propionic acid

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD08144104
CAS Number: 885273-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR