4-Methyl-2-piperidin-1-ylthiazole-5-carbaldehyde

Supplier: Matrix Scientific

021005-500MG
101811-172EA 297.85 USD
101811-172
4-Methyl-2-piperidin-1-ylthiazole-5-carbaldehyde
4-Methyl-2-piperidin-1-ylthiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₁₄N₂OS
MDL Number: MFCD04624838
CAS Number: 730997-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR