(2-Imidazol-1-ylethyl)methylamine dihydrochloride

Supplier: Matrix Scientific

020997-500MG
101811-156EA 209.3 USD
101811-156
(2-Imidazol-1-ylethyl)methylamine dihydrochloride
(2-Imidazol-1-ylethyl)methylamine dihydrochloride

Hazard: Irritant

Formula: C₆H₁₃Cl₂N₃
MDL Number: MFCD08687939
CAS Number: 106891-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR