1-Adamantan-1-ylpiperazine

Supplier: Matrix Scientific

020987-500MG
101811-146EA 52.9 USD
101811-146
1-Adamantan-1-ylpiperazine
1-Adamantan-1-ylpiperazine

Hazard: Irritant

Formula: C₁₄H₂₄N₂
MW: 220.36 g/mol
MDL Number: MFCD01748381
CAS Number: 19984-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR