4-Methyl-5-phenyl-2H-pyrazol-3-ylamine

Supplier: Matrix Scientific

020945-500MG
101811-064EA 280 USD
101811-064
4-Methyl-5-phenyl-2H-pyrazol-3-ylamine
4-Methyl-5-phenyl-2H-pyrazol-3-ylamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃
MDL Number: MFCD05864593
CAS Number: 66367-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR