2-[2-(2-Methylpiperidin-1-yl)ethoxy]benzaldehyde

Supplier: Matrix Scientific

020930-500MG
101811-038EA 280 USD
101811-038
2-[2-(2-Methylpiperidin-1-yl)ethoxy]benzaldehyde
2-[2-(2-Methylpiperidin-1-yl)ethoxy]benzaldehyde

Hazard: Irritant

Formula: C₁₅H₂₁NO₂
MW: 247.34 g/mol
MDL Number: MFCD08146607
CAS Number: 883542-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR