3,5-Dimethyl-1H-pyrrole-2,4-dicarboxylic acid-4-ethyl ester

Supplier: Matrix Scientific

020905-500MG
101810-986EA 28.75 USD
101810-986
3,5-Dimethyl-1H-pyrrole-2,4-dicarboxylic acid-4-ethyl ester
3,5-Dimethyl-1H-pyrrole-2,4-dicarboxylic acid-4-ethyl ester

Hazard: Irritant

Formula: C₁₀H₁₃NO₄
MDL Number: MFCD00030408
CAS Number: 5442-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR