2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(2-Phenylthiazol-4-yl)acetic acid

020903-500MG
101810-982EA 77.05 USD
101810-982
2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid
2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂S
MW: 219.26 g/mol
MDL Number: MFCD01312954
CAS Number: 16441-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR