1-Phenethyl-1H-benzoimidazol-5-ylamine dihydrochloride

Supplier: Matrix Scientific

020890-500MG
101810-958EA 280 USD
101810-958
1-Phenethyl-1H-benzoimidazol-5-ylamine dihydrochloride
1-Phenethyl-1H-benzoimidazol-5-ylamine dihydrochloride

Hazard: Irritant

Formula: C₁₅H₁₇Cl₂N₃
MDL Number: MFCD08688457
CAS Number: 1179363-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR