(2,4-Diethoxy-6-methylpyrimidin-5-yl)acetic acid

Supplier: Matrix Scientific

020887-500MG
101810-952EA 201.25 USD
101810-952
(2,4-Diethoxy-6-methylpyrimidin-5-yl)acetic acid
(2,4-Diethoxy-6-methylpyrimidin-5-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₄
MDL Number: MFCD07186333
CAS Number: 500159-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR