1-(3,5-Dimethylisoxazole-4-sulfonyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

020879-500MG
101810-938EA 209.3 USD
101810-938
1-(3,5-Dimethylisoxazole-4-sulfonyl)piperidine-4-carboxylic acid
1-(3,5-Dimethylisoxazole-4-sulfonyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₅S
MDL Number: MFCD07343993
CAS Number: 697258-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR