2-Amino-N-tert-butylacetamide hydrochloride

Supplier: Matrix Scientific

020878-500MG
101810-936EA 209.3 USD
101810-936
2-Amino-N-tert-butylacetamide hydrochloride
2-Amino-N-tert-butylacetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O
MDL Number: MFCD07324741
CAS Number: 71034-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR