1-(2-Hydroxy-3-pyrrolidin-1-yl-propyl)-2-methyl-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

020871-500MG
101810-922EA 209.3 USD
101810-922
1-(2-Hydroxy-3-pyrrolidin-1-yl-propyl)-2-methyl-1H-indole-3-carbaldehyde
1-(2-Hydroxy-3-pyrrolidin-1-yl-propyl)-2-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₂
MDL Number: MFCD08144882
CAS Number: 883543-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR