C-(4-Chlorophenyl)-C-pyridin-3-ylmethylamine

Supplier: Matrix Scientific

020857-500MG
101810-894EA 161 USD
101810-894
C-(4-Chlorophenyl)-C-pyridin-3-ylmethylamine
C-(4-Chlorophenyl)-C-pyridin-3-ylmethylamine

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂
MDL Number: MFCD08144803
CAS Number: 883548-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR