1-(4-Isopropylphenyl)ethylamine

Supplier: Matrix Scientific

020846-500MG
101810-880EA 370.3 USD
101810-880
1-(4-Isopropylphenyl)ethylamine
1-(4-Isopropylphenyl)ethylamine

Hazard: Irritant

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
MDL Number: MFCD02177306
CAS Number: 73441-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR