(Indan-5-yloxy)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-((2,3-Dihydro-1H-inden-5-yl)oxy)acetic acid

020845-500MG
101810-878EA 209.3 USD
101810-878
(Indan-5-yloxy)acetic acid
(Indan-5-yloxy)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
MDL Number: MFCD01672404
CAS Number: 1878-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR