2-Methyl-2-m-tolyloxypropionic acid

Supplier: Matrix Scientific

020844-500MG
101810-876EA 201.25 USD
101810-876
2-Methyl-2-m-tolyloxypropionic acid
2-Methyl-2-m-tolyloxypropionic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₃
MDL Number: MFCD01365790
CAS Number: 53498-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR