1-Pyrrolidin-1-ylmethylpropylamine

Supplier: Matrix Scientific

020835-500MG
101810-858EA 209.3 USD
101810-858
1-Pyrrolidin-1-ylmethylpropylamine
1-Pyrrolidin-1-ylmethylpropylamine

Hazard: Irritant

Formula: C₈H₁₈N₂
MDL Number: MFCD06446856
CAS Number: 883547-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR