1-(4-Chlorophenylsulfonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

012124-500MG
101801-958EA 148.35 USD
101801-958
1-(4-Chlorophenylsulfonyl)-2-pyrrolidinecarboxylic acid
1-(4-Chlorophenylsulphonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₄S
MDL Number: MFCD00172806
CAS Number: 73096-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR