1-Cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

Supplier: Matrix Scientific

012120-500MG
101801-950EA 82.8 USD
101801-950
1-Cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde
1-Cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₁₃NO
MDL Number: MFCD04309657
CAS Number: 640264-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR