4,5,6,7-Tetrahydro-1H-indazole-3-carboxylic acid

Supplier: Matrix Scientific

012071-500MG
101801-856EA 204.7 USD
101801-856
4,5,6,7-Tetrahydro-1H-indazole-3-carboxylic acid
4,5,6,7-Tetrahydro-1H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
Melting Pt: 254 °C
MDL Number: MFCD02939914
CAS Number: 6076-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR