3-(2-Oxopyrrolidin-1-yl)propionic acid

Supplier: Matrix Scientific

Synonyms: 3-(2-Oxopyrrolidin-1-yl)propanoic acid

012062-500MG
101801-838EA 141.45 USD
101801-838
3-(2-Oxopyrrolidin-1-yl)propionic acid
3-(2-Oxopyrrolidin-1-yl)propionic acid

Hazard: Irritant

Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
MDL Number: MFCD05237379
CAS Number: 77191-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR