(4-Oxo-3-quinazolinyl)acetic acid

Supplier: Matrix Scientific

012048-500MG
101801-812EA 204.7 USD
101801-812
(4-Oxo-3-quinazolinyl)acetic acid
(4-Oxo-3-quinazolinyl)acetic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₃
Melting Pt: 237…294 °C
MDL Number: MFCD02332713
CAS Number: 14663-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR