6-Fluoro-2-methylquinoline-4-carboxylic acid
Supplier: Matrix Scientific
011864-500MG
101801-468EA
125.35
USD
101801-468
6-Fluoro-2-methylquinoline-4-carboxylic acid
6-Fluoro-2-methylquinoline-4-carboxylic acid
CAS: 716-03-0
MDL Number: MFCD02741579
Molecular Formula: C11H8FNO2
Molecular Weight: 205,19
Hazard: Irritant
Formula:
C₁₁H₈FNO₂ Melting Pt: 246 °C |
MDL Number:
MFCD02741579 CAS Number: 716-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...