2-(4-Methylpiperazin-1-yl)benzoic acid
Supplier: Matrix Scientific
011682-500MG
101801-112EA
100.05
USD
101801-112
2-(4-Methylpiperazin-1-yl)benzoic acid
2-(4-Methylpiperazin-1-yl)benzoic acid
CAS: 159589-70-5
MDL Number: MFCD01788118
Molecular Formula: C12H16N2O2
Molecular Weight: 220,27
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ Melting Pt: 211…215 °C |
MDL Number:
MFCD01788118 CAS Number: 159589-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...