1-(4-Methoxybenzyl)piperazine hydrochloride

Supplier: Matrix Scientific

011642-500MG
101801-042EA 204.7 USD
101801-042
1-(4-Methoxybenzyl)piperazine hydrochloride
1-(4-Methoxybenzyl)piperazine hydrochloride

CAS: 109188-09-2
MDL Number: MFCD05656277
Molecular Formula: C12H19ClN2O
Molecular Weight: 242,75
Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD05656277
CAS Number: 109188-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR