3-Chlorobenzo[b]thiophene-2-carboxylic acid
011634-500MG
101801-026EA
936.48
USD
101801-026
3-Chlorobenzo[b]thiophene-2-carboxylic acid
3-Chlorobenzo[b]thiophene-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅ClO₂S MW: 212.65 g/mol Melting Pt: 263 °C |
MDL Number:
MFCD00014579 CAS Number: 21211-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...