2-{[(4-Fluorophenyl)sulfonyl]amino}-3-methylbutanoic acid

Supplier: Matrix Scientific

Synonyms: 2-(4-Fluorobenzenesulphamido)-3-methylbutyric acid

011630-500MG
101801-018EA 204.7 USD
101801-018
2-{[(4-Fluorophenyl)sulfonyl]amino}-3-methylbutanoic acid
2-{[(4-Fluorophenyl)sulfonyl]amino}-3-methylbutanoic acid

CAS: 250714-79-5
MDL Number: MFCD00664351
Molecular Formula: C11H14FNO4S
Molecular Weight: 275,3
Hazard: Irritant

Formula: C₁₁H₁₄FNO₄S
MW: 275.3 g/mol
MDL Number: MFCD00664351
CAS Number: 250714-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR